Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Iron(III) hexacyanoferrate(II), pure

Supplier: Thermo Scientific Chemicals
Synonyms: Prussian BlueIron(III) Ferrocyanide

21521-0250 21521-1000
TS21521-1000EA 166.93 USD InStock
TS21521-1000 TS21521-0250
Iron(III) hexacyanoferrate(II), pure
Iron(III) hexacyanoferrate(II)


Formula: C₁₈Fe₇N₁₈
MW: 859.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00135663
CAS Number: 14038-43-8
Order Now
Specification Test Results
Appearance Dark blue Powder
Infrared spectrum Conforms
UV spectrum Complies

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty