Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(cyclopentadienyl)titanium dichloride 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Titanocene dichlorideBis(cyclopentadienyl)titanium(IV) dichlorideTitanocenedichloride

21576-0100 21576-0500
TS21576-0500EA 170 USD InStock
TS21576-0500 TS21576-0100
Bis(cyclopentadienyl)titanium dichloride 97%
Bis(cyclopentadienyl)titanium dichloride


Formula: C₁₀H₁₀Cl₂Ti
MW: 248.97 g/mol
Melting Pt: 287…289 °C
Storage Temperature: Ambient
MDL Number: MFCD00003723
CAS Number: 1271-19-8
EINECS: 215-035-9
Order Now
Specification Test Results
Appearance Red to bordeaux Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty