Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(pentafluorophenyl)phosphine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tris(perfluorophenyl)phosphinePerfluorotriphenylphosphine

31316-0010 31316-0050
TS31316-0050EA 183 USD InStock
TS31316-0050 TS31316-0010
Tris(pentafluorophenyl)phosphine 97%
Tris(pentafluorophenyl)phosphine


Formula: C₁₈F₁₅P
MW: 532.15 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD00079654
CAS Number: 1259-35-4
EINECS: 215-021-2
Order Now
Specification Test Results
Appearance White to beige to light brown Powder or crystals
Infrared spectrum Conforms
Melting point 108 to 119 °C
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty