Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ninhydrin for TLC

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2,2-Dihydroxyindan-1,3-dionTrioxohydrindene monohydrateNinhydrin monohydrate2,2-Dihydroxy-1,3-indanedioneIndanetrione hydrate2,2-Dihydroxyindane-1,3-dione1,2,3-Indantrione monohydrate

32702-2400
TS32702-2400EA 125 USD InStock
TS32702-2400
Ninhydrin for TLC
Ninhydrin


Formula: C₉H₆O₄
MW: 178.14 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 250…258 °C
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
UN: 1950
Merck Index: 1206646
https://us-legacy.vwr.com/stibo/web/std.lang.all/70/01/2297001.jpg
Order Now
Specification Test Results
Additional info TLC spray reagent at 0.5 % in 1-Butanol
Additional info Ozone friendly

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty