Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triphenylphosphine 1.2 - 1.5 mmol/g on resin, cross-linked with 1% DVB 200-400 mesh

Supplier: Thermo Scientific Chemicals
Warning

36684-0050
TS36684-0050EA 98 USD InStock
TS36684-0050
Triphenylphosphine 1.2 - 1.5 mmol/g on resin, cross-linked with 1% DVB 200-400 mesh
Triphenylphosphine


Formula: C₁₈H₁₅P
MW: 262.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00003043
CAS Number: 603-35-0
Order Now
Specification Test Results
Appearance White to yellow Beads
Particle size 200 to 400 mesh
Crosslinking 1 % DVB
Loading 1 to 1.5 mmol/g

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty