Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Povidone iodine

Supplier: Thermo Scientific Chemicals
Synonyms: Polyvinylpyrrolidone-iodine complex

22947-1000 22947-0025 22947-5000
TS22947-5000EA 332 USD InStock
TS22947-5000 TS22947-1000 TS22947-0025
Povidone iodine
Povidone iodine


Formula: C₆H₉I₂NO
MW: 364.953 g/mol
Melting Pt: 300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00084483
CAS Number: 25655-41-8
Order Now
Specification Test Results
Appearance Yellow to red-brown Powder
Infrared spectrum Conforms
Assay 9.0 to 12.0 % (available iodine) (on dried substance)
Loss on drying ≤8 % (5 g, 105°C)
Sulfated ash ≤0.025 % (2 g)
Iodide (I) ≤6.6 % (on dried substance)
Nitrogen (N) 9.5 to 11.5 % (on dried substance)
Heavy metals (as Pb) ≤20 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty