Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabromomethane 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Carbon tetrabromide

32515-5000 32515-1000 32515-0050
TS32515-1000EA 80.16 USD InStock
TS32515-1000 TS32515-0050 TS32515-5000
Tetrabromomethane 98%
Tetrabromomethane


Formula: Br₄C
MW: 331.63 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: 88…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00000117
CAS Number: 558-13-4
EINECS: 209-189-6
UN: 2516
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to off-white Crystalline solid
Water ≤10 % (K.F.)
GC ≥97.5 % (On organic basis)
Stabilizer May contain water as stabilizer

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty