Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorodiphenylphosphine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: DiphenylchlorophosphineDPPC

34730-5000 34730-0250 34730-0050 34730-1000
TS34730-0050EA 26.82 USD InStock
TS34730-0050 TS34730-5000 TS34730-0250 TS34730-1000
Chlorodiphenylphosphine 98%
Chlorodiphenylphosphine


Formula: C₁₂H₁₀ClP
MW: 220.64 g/mol
Boiling Pt: 320 °C (760 mmHg)
Density: 1.229 g/cm³
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00000529
CAS Number: 1079-66-9
EINECS: 214-093-2
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to yellow Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
Refractive index 1.6350 to 1.6370 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty