Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloro(diethyl)phosphine 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Diethylchlorophosphine

35031-0010 35031-0050
TS35031-0050EA 330 USD InStock
TS35031-0050 TS35031-0010
Chloro(diethyl)phosphine 95%
Chloro(diethyl)phosphine


Formula: C₄H₁₀ClP
MW: 124.55 g/mol
Boiling Pt: 131…132 °C (760 mmHg)
Density: 1.023 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00013621
CAS Number: 686-69-1
EINECS: 211-689-4
UN: 3129
ADR: 4.3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4760 (20°C, 589 nm)
NMR ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty