Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

O-Benzylhydroxylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

38205-1000 38205-0250 38205-0050
TS38205-0050EA 74 USD InStock
TS38205-0050 TS38205-0250 TS38205-1000
O-Benzylhydroxylamine 97%
O-Benzylhydroxylamine


Formula: C₇H₉NO
Boiling Pt: 110…112 °C (20 mmHg)
Density: 1.19 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00221709
CAS Number: 622-33-3
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5390 to 1.5410 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty