Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ishikawa's reagent 90%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-(Diethylamino)-1,1,2,3,3,3-hexafluoropropaneN,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamineHexafluoropropene diethylamine

38921-1000 38921-0250
TS38921-0250EA 74 USD InStock
TS38921-0250 TS38921-1000
Ishikawa's reagent 90%
Ishikawa's reagent


Formula: C₇H₁₁F₆N
MW: 223.16 g/mol
Density: 1.23 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00054683
CAS Number: 309-88-6
UN: 2924
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC 90 % (Sum of two isomers) Approximate composition

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty