Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Aminofluorescein (isomer I), pure

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-oneFluoresceinamine, isomer I5-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

40077-0010 40077-0050
TS40077-0050EA 297 USD InStock
TS40077-0050 TS40077-0010
5-Aminofluorescein (isomer I), pure
5-Aminofluorescein (isomer I)


Formula: C₂₀H₁₃NO₅
MW: 347.33 g/mol
Melting Pt: 223 °C
Storage Temperature: Ambient
MDL Number: MFCD00005052
CAS Number: 3326-34-9
Order Now
Specification Test Results
Appearance Dark red-brown Crystalline powder
Infrared spectrum Conforms
HPLC 95 % Typical
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty