Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromothymol blue sodium salt ACS

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonateSodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulfonateSodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolateBromothymol blue water soluble

40326-0100
TS40326-0100EA 61 USD InStock
TS40326-0100
Bromothymol blue sodium salt ACS
Bromothymol blue sodium salt


Formula: C₂₇H₂₇Br₂NaO₅S
MW: 646.37 g/mol
Melting Pt: > 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00077263
CAS Number: 34722-90-2
EINECS: 252-169-7
Order Now
Specification Test Results
Appearance Dark green to orange-brown Powder
Infrared spectrum Conforms
Visual transition interval from pH 6.0 (yellow) to pH 7.6 (blue)
Clarity of solution Passes test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty