Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl violet 80%

Supplier: Thermo Scientific Chemicals
Synonyms: Basic Violet 4

41049-0250
TS41049-0250EA 42 USD InStock
TS41049-0250
Ethyl violet 80%
Ethyl violet


Formula: C₃₁H₄₂ClN₃
MW: 560.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011765
CAS Number: 2390-59-2
Order Now
Specification Test Results
Appearance Green Powder
Infrared spectrum Conforms
Loss on drying ≤4 %
Dye content ≥75 %
UV Lambda max 593 to 599 nm (c=0.1, H2O)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty