Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenolphthalein ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

41718-0026 41718-1000 41718-5000
TS41718-0026EA 418 USD InStock
TS41718-0026 TS41718-1000 TS41718-5000
Phenolphthalein ACS
Phenolphthalein


Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Melting Pt: 258…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 13,07325
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/01/2297201.jpg
Order Now
Specification Test Results
Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Visual transition interval From pH 8.0 (colorless) to pH 10 (red)
Clarity of alcohol solution Passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty