Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rhodamine 6G 99%, pure laser grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride

41902-5000 41902-0010 41902-0050
TS41902-0010EA 126 USD InStock
TS41902-0010 TS41902-0050 TS41902-5000
Rhodamine 6G 99%, pure laser grade
Rhodamine 6G


Formula: C₂₈H₃₁ClN₂O₃
MW: 479.02 g/mol
Melting Pt: 229.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00012665
CAS Number: 989-38-8
EINECS: 213-584-9
UN: 2811
Order Now
Specification Test Results
Appearance Red-brown to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
UV Lambda max 527 to 533 nm (in methanol)
Suitability for use as a laser dye
Molar extinction coefficient ≥105000 (lambda max)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty