Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

4-(Dimethylamino)benzaldehyde ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ehrlich's Reagent4-(N,N-Dimethylamino)benzaldehydep-Dimethylamino-Benzaldehydep-(Dimethylamino)benzaldehyde

42363-0025 42363-1000 42363-0250 42363-5000
TS42363-0025EA 1057 USD InStock
TS42363-0025 TS42363-1000 TS42363-0250 TS42363-5000
4-(Dimethylamino)benzaldehyde ACS
4-(Dimethylamino)benzaldehyde


Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: 176…177 °C (17 mmHg)
Melting Pt: 70…75 °C
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
EINECS: 202-819-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/62/54/746254.jpg
Order Now
Specification Test Results
Appearance Light yellow to light green Crystalline powder
Infrared spectrum Conforms
Melting point 73 to 75 °C
Solubility (in alcohol) Passes test, (in hydrochloric acid) Passes test
Residue after ignition ≤0.1 %
Color of alcohol solution ≤60 APHA
Color of a HCl solution Passes test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty