Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dess-Martin periodinane 15% (w/w) in dichloromethane, AcroSeal™

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl esterDMP1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one(1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one

42900-1000
TS42900-1000EA 359 USD InStock
TS42900-1000
Dess-Martin periodinane 15% (w/w) in dichloromethane, AcroSeal™
Dess-Martin periodinane


Formula: C₁₃H₁₃IO₈
MW: 424.15 g/mol
Density: 1.362 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00130127
CAS Number: 87413-09-0
UN: 1593
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Colorless to light yellow Solution (May develop some turbidity or precipitate)
Titration Iodometric 10 to 15 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty