Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Toluenesulfonyl isocyanate 96%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-IsocyanatosulphonyltolueneTosyl isocyanate4-Isocyanatosulfonyltoluene

43076-1000
TS43076-1000EA 40.64 USD InStock
TS43076-1000
p-Toluenesulfonyl isocyanate 96%, AcroSeal™
p-Toluenesulphonyl isocyanate


Formula: C₈H₇NO₃S
MW: 197.21 g/mol
Boiling Pt: 144 °C (10 mmHg)
Melting Pt: –2 °C
Density: 1.29 g/cm³
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00002030
CAS Number: 4083-64-1
UN: 2206
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5330 to 1.5360 (20°C, 589 nm)
GC ≥95 %
Color scale ≤50 APHA
Impurity ≤1.1 % Tosyl chloride

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty