Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylacrylamide 99% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 500 ppm

43202-1000 43202-5000
TS43202-1000EA 71.25 USD InStock
TS43202-1000 TS43202-5000
N,N-Dimethylacrylamide 99% stabilized
N,N-Dimethylacrylamide


Formula: C₅H₉NO
Boiling Pt: 80 °C (20 mmHg)
Density: 0.962 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008626
CAS Number: 2680-03-7
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4730 to 1.4750 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Approx. 500 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty