Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trichloroacetic anhydride 95%

Supplier: Thermo Scientific Chemicals
Danger

44609-1000 44609-0250
TS44609-0250EA 104 USD InStock
TS44609-0250 TS44609-1000
Trichloroacetic anhydride 95%
Trichloroacetic anhydride


Formula: C₄Cl₆O₃
MW: 308.76 g/mol
Boiling Pt: 139…141 °C (60 mmHg)
Melting Pt: –2 °C
Density: 1.69 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000793
CAS Number: 4124-31-6
UN: 3265
ADR: 8,I
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4870 (20°C, 589 nm)
Titration by coupling ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty