Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diisopropylcarbodiimide 99%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N,N'-methanetetraylbis(1-methylethylamine)1,3-DiisopropylcarbodiimideN,N-DiisopropylcarbodiimideN,N'-DiisopropylcarbodiimideN.N’-DiisopropylcarbodiimideDIC

44618-1000
TS44618-1000EA 212 USD InStock
TS44618-1000
Diisopropylcarbodiimide 99%, AcroSeal™
Diisopropylcarbodiimide


Formula: C₇H₁₄N₂
MW: 126.2 g/mol
Boiling Pt: 145…148 °C (760 mmHg)
Density: 0.81 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00065689
CAS Number: 693-13-0
UN: 3384
ADR: 6.1,
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4320 to 1.4350 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty