Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perfluorokerosene 50% in PFK low boiling, high boiling for mass spectrometry

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: PFK

45013-0010
TS45013-0010EA 336 USD InStock
TS45013-0010
Perfluorokerosene 50% in PFK low boiling, high boiling for mass spectrometry
Perfluorokerosene


Boiling Pt: 180 °C
Density: 1.89 g/cm³
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00132835
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Suitability (for mass analysis) Passes test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty