Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water cell culture reagent

Supplier: LONZA WALKERSVILLE INC
Synonyms: Dihydrogen oxide

17-724F 17-724F 17-724Q 17-724Q
12001-356EA 18.37 USD InStock
CA12001-706 CA12001-356 12001-356 12001-706
Water cell culture reagent
Water
Water for Cell Culture,Water For Injection (WFI) quality water is prepared by ultrafiltration, reveresed osmosis, deionizaton and sterile filtration.

Lonza’s Water for Cell Culture is aseptically processed from a validated Water for Injection (WFI) system that meets current USP specifications.


Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty