Would you like to log in again?
CryoCube® F440n ULT Freezers
Supplier: Eppendorf
CryoCube®
F440202035 F440203035 F440200035 F440202035 F440200035 F440203035
76542-106EA 18991.63 USD InStock
76542-106 CA76542-104 76542-104 CA76542-108 CA76542-106 76542-108
CryoCube® F440n ULT Freezers
Freezers Ultra Low Temperature Freezers
CryoCube® F440n offers a 5-compartment lay-out and 5 insulated inner doors for easy sample access and safe sample storage at −80 °C. The walls are 130 mm thick based on foam, it has a volume of 440 L (15.5 cubic feet) and a temperature range from −50 to −86 °C.
  • 130 mm wall (polyurethane insulation)
  • 'Green' hydrocarbon-based cooling liquids (R290/R170)
  • Air-cooling
These ULT freezers are equipped with reliable heavy-duty compressors (2-stage cascade cooling system). Quick pull-down times enable fast re-use after cleaning/defrosting of the instrument. The freezers have short recovery times after opening to provide extra sample safety (Door-Open-Recovery/DOR) when getting back to −80 °C.
The automatic vent port enables fast re-access if needed. For even higher sample safety, the ULT freezers can be equipped with CO₂/LN₂ back-up systems and a chart recorder. Temperature monitor systems can be used to monitor the freezers remotely.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty