Would you like to log in again?
HardyCHROM™ Group A Strep Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
G337
76499-316PK 93.71 USD InStock
76499-316
HardyCHROM™ Group A Strep Agar, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
HardyCHROM™ Group A Strep agar is a chromogenic medium for the selective cultivation and differential isolation of Group A Streptococcus (S. pyogenes) from specimens based on colony color.
  • Selects for Strep Group A (Streptococcus pyogenes)
  • Red colonies stand out on a white background for easy detection
The HardyCHROM™ plates use optically clear, heavy-duty polystyrene mono plates for distortion-free viewing and reduced breakage during shipping. Receive your pack in a 'breathable' sleeve of 10 plates per bag that prevents the buildup of condensation and excess moisture. Hardy mono plates feature raised rings for stacking ability and prevent sliding.
Packaging: 10 per pack.
Caution: This media is to be stored and incubated in the dark.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty