Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Rubbermaid® TC Microburst 9000 Air Freshener Refill, Mandarin Orange
Supplier: Essendant
RCP402093
76540-120CT 80.55 USD InStock
76540-120
Rubbermaid® TC Microburst 9000 Air Freshener Refill, Mandarin Orange
Deodorants and Air Fresheners
Refreshes any washroom with an invigorating scent. This commercial air freshener refill offers 90 to 180 days of maximum odor control distributed in a fine dry mist with 63% fewer VOCs per spray than standard aerosol refills.
  • Refill actuator dispenses a fine mist of long lasting fragrance
  • Effectively controls odors for 90 to 180 days
  • Invigorating Mandarin Orange fragrance
  • For use with Rubbermaid Commercial Microburst 9000 dispensers (sold separately)
This aerosol spray refill is designe for use with the Rubbermaid Commercial Microburst 9000 Aerosol air freshener dispenser (sold separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty