Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hammermill® Colors Printer Paper, Gray, 20 lbs. Bond Weight

Supplier: Essendant
Hammermill®
HAM102889
76540-084PK 13.62 USD InStock
76540-084
Hammermill® Colors Printer Paper, Gray, 20 lbs. Bond Weight
Paper
High-quality Colors printer paper increases the visual appeal of documents to help them stand out from the crowd.

  • Acid-free for archival quality
  • Smooth, even surface with a high opacity for minimal show-through
  • Contributes toward satisfying credit MR1 under LEED
  • Paper basis weight: 20 lbs. bond
  • Paper grade: 75 gsm

Dependable and efficient, it runs smoothly through all equipment. Precise, accurate sheeting, consistent moisture content, and curl control ensure smooth, trouble-free performance. Good opacity to minimize show-through. Excellent for color-coding. Backed by the 99.99% Jam-Free Guarantee. Acid-free material prevents yellowing over time to ensure a long-lasting appearance.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty