Would you like to log in again?
Masterflex® Hose Clamps, Nylon, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
MAKEUP MAKEUP MAKEUP MAKEUP MAKEUP MAKEUP MAKEUP MAKEUP MAKEUP MAKEUP
MFLX98766-31EA 38.8 USD InStock
MFLX98766-31 MFLX98766-42 MFLX98766-32 MFLX98766-33 MFLX98766-34 MFLX98766-35 MFLX98766-36 MFLX98766-37 MFLX98766-38 MFLX98766-41
Masterflex® Hose Clamps, Nylon, Avantor®
Clamps Tubing and Sanitary Clamps
Secure tubing to connectors, pumps, glassware, and filtration devices.
  • Use to fasten pressure, vacuum, and liquid tubing lines
  • Nylon is corrosion-proof, chemical resistant, and non-conductive
  • Works with all types of plastic and rubber tubing
Reusable clamp squeezes shut for a leakproof seal—press the sides to release the clamp. Order the size that suits your needs, or the handy kit to handle multiple projects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty