Would you like to log in again?
Masterflex® B/T® Fixed Speed Analog Drives, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
77111-30 77111-37
MFLX77111-30EA 7280 USD InStock
MFLX77111-30 MFLX77111-37
Masterflex® B/T® Fixed Speed Analog Drives, Avantor®
Pumps Peristaltic Pumps
High-volume transfer and dispensing.
  • Flow rate 18.9/16 or 42/35 LPM with B/T tubing—flow rate depends on drive and tubing size
  • Rapid-Load® pump head occlusion mechanism slides occlusion bed out of the way for easy tube loading and improved tubing occlusion
  • Accepts B/T PerfectPosition™ pump tubing sizes B/T 87 and B/T 91
  • Interlock switch shuts off power to stop drive when door is opened
Masterflex® B/T® drives are the best choice for high-flow bulk-transfer applications. Their durable construction ensures long service life with minimal maintenance. The sturdy materials of construction include stainless steel/carbon steel, a cast aluminum housing with an epoxy-powder coating, and Nylatron® rollers for rugged dependability.
All B/T drives include a new and improved Rapid-Load® pump head for fast and easy tubing changes. The redesigned occlusion mechanism provides an improved tubing retention mechanism which holds the tubing firmly in place to further simplify the loading process. PerfectPosition™ tubing sizes provide improved performance in higher-pressure applications and provide longer tubing life.
Delivery information: Includes rapid-load pump head, 3 ft (1 m) of Tygon LFL B/T 91 tubing 06430-91, and 6-ft (1.8 m) line cord with IEC 320/CEE22 socket; pump is shipped with country-specific plug/cord set.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty