Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Masterflex® Adapter Fittings, Sanitary Clamp to Threaded, Straight, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
3070072 3070076 3070066 3070078 3070068 3070079 3070071 3070069 3070077 3070074 3070067 3070070
MFLX30700-76EA 144 USD InStock
MFLX30700-76 MFLX30700-74 MFLX30700-72 MFLX30700-71 MFLX30700-70 MFLX30700-69 MFLX30700-68 MFLX30700-79 MFLX30700-78 MFLX30700-67 MFLX30700-66 MFLX30700-77
Masterflex® Adapter Fittings, Sanitary Clamp to Threaded, Straight, Avantor®
Fittings and Connectors
Meet FDA requirements and 3A Sanitary Standards.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Compatible with standard 304L fittings clamps
These threaded adapter clamps by NPT(M) sanitary fittings are ideal for quick and easily installation of your sanitary set-up to an NPT male pipe thread. The 316L stainless steel couplings are suitable for the pharmaceutical industry. They offer great chemical compatibility for use with corrosive acids and bases. Clamps and gaskets required for a complete system are sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty