Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Treston® Cornerstone Lab Benches

Supplier: Treston
14-CS918-49 14-CS924-49 14-CS926-49 14-CS922-49 14-CS919-49 14-CS921-49 14-CS925-49 14-CS901-49 14-CS909-49 14-CS917-49 14-CS910-49 14-CS928-49 14-CS912-49 14-CS905-49 14-CS923-49 14-CS911-49 14-CS906-49 14-CS927-49 14-CS920-49 14-CS908-49 14-CS907-49 14-CS903-49 14-CS902-49 14-CS904-49
76532-894EA 2153.37 USD InStock
76532-894 76532-930 76532-920 76532-898 76532-940 76532-896 76532-918 76532-908 76532-934 76532-912 76532-902 76532-924 76532-932 76532-910 76532-922 76532-900 76532-916 76532-938 76532-928 76532-906 76532-936 76532-914 76532-926 76532-904
Treston® Cornerstone Lab Benches
Furniture Tables and Workbenches
Treston® Cornerstone lab benches are manually height adjustable in 1" increments. All bench frames can be equipped with casters to create a mobile workstation solution.

  • Heavy-duty steel structure supports loads up to 2000 lbs.
  • A cost-effective option
  • Upright tubes allow for attachment of a wide range of accessories
  • Light grey frame color

Treston® lab benches are ergonomically designed to meet the needs of laboratory personnel. There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality.

The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress.

Accessories information: Suitable for use with Treston® workstation accessories.

Ordering information: Available with laminate or black phenolic worksurfaces. Five-year warranty on all Treston® steel products. Electric components have a two-year warranty. Accessories not included unless otherwise noted. Installation is required. Assembly videos available.

Delivery information: Benches ship LTL on pallets for delivery. When ordering, please specify any special delivery requirements.

Packaging: Benches ship unassembled. Assembled shipping and installation services available upon request.

Caution: Lab bench frames and tops can be heavy and may require two-man lift.

Order Now
 

ORDER

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty