Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Masterflex® L/S® High-Pressure Pump Tubing, PharMed® BPT, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
AXB42848-CP-1FO AXB42241-CP-1FO
MFLX05664-24CS 0 USD InStock
MFLX05664-24 MFLX05664-16
Masterflex® L/S® High-Pressure Pump Tubing, PharMed® BPT, Avantor®
Tubing Peristaltic Pump Tubing
Designed to pump at higher pressures.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples
Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige
High-Pressure PharMed® BPT tubing is designed for use with the L/S® High-Performance pump head models 77250-62 and 77250-82 only.
Please note that the maximum drive speed for these High-Pressure PharMed® BPT tubing is 100 RPM.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty