Would you like to log in again?
Masterflex® Polypropylene Bag Filter Housings, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
150360 150338 150337 150367
MFLX01510-00EA 321 USD InStock
MFLX01510-00 MFLX01510-06 MFLX01510-04 MFLX01510-02
Masterflex® Polypropylene Bag Filter Housings, Avantor®
Filters Bag Filter Systems
Strength and durability without bulk.
  • Polypropylene construction provides corrosion resistance, strength, and durability
  • Housings are lightweight and portable
  • Maximum temperature is 100 °F (38 °C)
Polypropylene is ideal for water, acids, plating solutions, corrosive chemicals, food products, and more. Each housing includes a Buna-N O-ring, gauge, wrench, ¼" vent port with plug, and 3/8" drain valve.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty