Would you like to log in again?
Masterflex® Union Fittings, Female NPT Threaded, Elbow, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
MAKE-UP PTE-14 (6PK) MAKE-UP MAKE-UP MAKE-UP
MFLX06349-60EA 33.25 USD InStock
MFLX06349-60 MFLX06349-61 MFLX06349-64 MFLX06349-62 MFLX06349-63
Masterflex® Union Fittings, Female NPT Threaded, Elbow, Avantor®
Fittings and Connectors
Chemically-inert PFA handles most aggressive chemicals.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings offer a more permanent installation. Choose the size to meet your application needs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty