Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Silica Vacu-vials Instrumental Kit, CHEMetrics
Supplier: CHEMetrics
K-9003
CXK-9003EA 50.97 USD InStock
CXK-9003
Silica Vacu-vials Instrumental Kit, CHEMetrics
Environmental Test Kits
An instrumental test kit for the colorimetric analysis of silica in water. This test kit utilizes Vacu-vials® self-filling ampoules.
  • Heteropoly Blue Method
The silica content of water should be determined prior to its use in industrial applications as silica can form a harmful scale on equipment and heat transfer surfaces, particularly steam turbine blades. Silica will also foul reverse osmosis membranes reducing efficiency. Silica is often tested in boiler water, cooling water, natural water, wastewater, and high purity water environments.
Ordering information: This kit requires the use of a CHEMetrics Direct-Readout Photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.
Delivery information: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, Neutralizer Solution, Activator Solution, 25 mL sample cup, sample cup top, ampoule blank, and instructions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty