Would you like to log in again?
Masterflex® Storage Racks for Fitting Kits, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
EJK143 LLG STORAGE RACK EJK142
MFLX31513-12EA 88.8 USD InStock
MFLX31513-12 MFLX31513-32
Masterflex® Storage Racks for Fitting Kits, Avantor®
Boxes Storage Boxes
Organize fittings for easy access.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Organize your workspace with these Masterflex® fitting kit storage racks. Storage racks hold up to four fitting kits. Simply slide the complete fitting kit into the rack for secure storage. Storage racks are for kits only, not assortment packs. Fitting kit with fittings are sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty