Would you like to log in again?
Masterflex® Transfer Tubing, Convoluted Perfluroalkoxy, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
7446
MFLX06487-04EA 202 USD InStock
MFLX06487-04
Masterflex® Transfer Tubing, Convoluted Perfluroalkoxy, Avantor®
Tubing Transfer Tubing
Promotes full drainage.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Convoluted spiral design promotes full drainage and increases tubing flexibility. This chemical-resistant fluoropolymer tubing is sold in 5-ft lengths with nonconvoluted 1"L sections at both ends for easy connections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty