Would you like to log in again?
Masterflex® Transfer Tubing, Coiled FEP, 12" Relaxed, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
49125 46034 34-0007-01
MFLX95821-04EA 335 USD InStock
MFLX95821-04 MFLX95821-02 MFLX95821-00
Masterflex® Transfer Tubing, Coiled FEP, 12" Relaxed, Avantor®
Tubing Transfer Tubing
Retracts for easier handling.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Our retractable coil FEP tubing offers the chemical inertness of our standard FEP tubing combined with the flexibility of a coil. FEP tubing has excellent heat resistance, a low coefficient of friction, and won't deteriorate with age. Tubing withstands temperatures from -454 to 400 °F (-270 to 205 °C); and can be sterilized by autoclave, with ethylene oxide, or by using dry heat. Choose the size and length that best fits your application. Use tubing with compression fittings.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty