Would you like to log in again?
Masterflex® I/P® Fixed-Speed Wash-Down Drives, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
7588-60 7588-30 7588-35
MFLX07588-60EA 3340 USD InStock
MFLX07588-60 MFLX07588-30 MFLX07588-35
Masterflex® I/P® Fixed-Speed Wash-Down Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Easy operation and easy
on the budget.
  • Purge system or pump in either direction with combination on/off/reverse switch
  • Constructed of painted steel/aluminum IP55-rated housing
  • Get more flow—capable of driving multiple pump heads for multichannel pumping
These fixed-speed drives are IP55-rated for protection from dust and water. Accept I/P standard, Easy-Load®, or High-Performance pump heads. Stack up to two Standard or Easy-Load pump heads to double flow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty