Would you like to log in again?
Masterflex® High-Purity Filter Cartridge Housings, Polypropylene, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
150134
MFLX01508-24EA 115 USD InStock
MFLX01508-24
Masterflex® High-Purity Filter Cartridge Housings, Polypropylene, Avantor®
Filters Filter Cartridges
Filter housing is ideal for in-line process filtration systems.
  • Constructed of durable polypropylene (PP)
This #12 housing fits submicronic cartridges with 222 O-ring sealing caps. The pure PP housing contains no filters, lubricants, or plasticizers. Housing features an O-ring sealed PP drain plug in the sump with a second vent for differential pressure.
Maximum temperature is 100 °F (38 °C); maximum pressure is 100 psi. Inlet/outlet ports are 3/4" NPT(F); length is 14-3/8". Includes VITON O-ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty