Would you like to log in again?
Masterflex® Adapter Fittings, Compression to Female Threaded, Straight, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP
MFLX06382-62EA 32 USD InStock
MFLX06382-62 MFLX30803-48 MFLX30803-49 MFLX30805-48 MFLX30805-49 MFLX30803-47 MFLX06384-60 MFLX30805-47 MFLX06384-40 MFLX06384-64 MFLX06384-20 MFLX30815-57 MFLX30815-56 MFLX30815-53 MFLX30815-52 MFLX30815-55 MFLX30815-54 MFLX06384-10 MFLX06382-59 MFLX06382-58 MFLX06382-57 MFLX06382-56 MFLX06384-50 MFLX06384-30 MFLX30805-51 MFLX30805-52 MFLX30805-50
Masterflex® Adapter Fittings, Compression to Female Threaded, Straight, Avantor®
Fittings and Connectors
Maintain a permanent connection with these adapters.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These threaded fittings are suitable for permanent installation and can used with your rigid tubing for high-pressure applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty