Would you like to log in again?
Linearity FLQ HbA1c for Bio-Rad Systems
Supplier: AUDIT Microcontrols
K761M-5 K983M-5
77391-858EA 480.77 USD InStock
77391-858 76545-590
Linearity FLQ HbA1c for Bio-Rad Systems
Control reagents
Linearity FLQ HbA1c for Bio-Rad® Systems is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for HbA1c.
  • Linearity; Frozen Liquid
The Audit® MicroControlsTM Linearity FLQ HbA1c is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analyte: HbA1c.
Delivery information: Priority overnight.
Packaging: Kits are shrinked-wrapped.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty