Would you like to log in again?
Desalkylflurazepam (CRM), Cayman Chemical
Supplier: Cayman Chemical
19698-1
76544-118EA 37.7 USD InStock
76544-118
Desalkylflurazepam (CRM), Cayman Chemical
Standards Organic Standards
An active metabolite of several benzodiazepines that inhibits L-type Cav channels (IC50s = 55 and 37 µM for Cav1.2 and 1.3, respectively) by positively modulating GABAA receptors
  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty