Would you like to log in again?
J&W CP-TAP CB Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
CP7483 CP7483
AGCP7483EA 1206 USD InStock
AGCP7483 CAAGCP7483
J&W CP-TAP CB Columns, Agilent Technologies
Chromatography Columns
Engineered for the detailed analysis of triglycerides based on carbon number and degree of unsaturation.
  • Engineered phase for detailed analysis of triglycerides
  • Separation based on carbon number and degree of unsaturation
  • Stabilized phase for low bleed and enhanced column lifetime
  • Available in Fused Silica and UltiMetal
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W CP-TAP CB for Triglycerides provides resolution based on carbon number and degree of unsaturation, delivering a more refined separation of complete triglyceride pattern in less than 16 minutes. The chemically-bonded phase exhibits low bleed and provides longer column lifetimes. The column is available in special fused silica for maximum column strength at temperatures up to 370 °C, or UltiMetal stainless steel capillary for the ultimate robustness.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty