Would you like to log in again?
J&W CP-TCEP for Alcohols in Gasoline Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
CP7525 CP7525
AGCP7525EA 1582 USD InStock
CAAGCP7525 AGCP7525
J&W CP-TCEP for Alcohols in Gasoline Columns, Agilent Technologies
Chromatography Columns
A highly polar column that separates benzene after n-dodecane, ideal for analysis of alcohols and aromatics in gasoline.
  • Engineered for analysis of alcohols in gasoline
  • Excellent peak shape for accurate separation of alcohols and aromatics in gasoline
  • Temperature stable to 135 °C for high productivity
  • Unique selectivity separates benzene after n-dodecane
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W CP-TCEP for Alcohols in Gasoline is a highly polar phase, engineered for the analysis of alcohols and aromatics in gasoline. It is able to elute aliphatic compounds smaller than n-dodecane before benzene, avoiding confusing aliphatic and aromatic fractions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty