Would you like to log in again?
J&W DB-MTBE Columns, Agilent Technologies
Supplier: Agilent Technologies
J&W
124-0034 124-0034
AG124-0034EA 926 USD InStock
CAAG124-0034 AG124-0034
J&W DB-MTBE Columns, Agilent Technologies
Chromatography Columns
A low-polarity stationary phase column that resolves MTBE from 2-methylpentane and 3-methylpentane.
  • Low-polarity stationary phase
  • Resolves MTBE from 2-methylpentane and 3-methylpentane for more accurate quantitation
  • Engineered for purge-and-trap injection without the need for cryofocusing
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-MTBE columns feature a low-polarity stationary phase that resolves MTBE from 2-methylpentane and 3-methylpentane. Extensively tested to the strictest industry QC standards, these columns are bonded, crosslinked, and solvent rinsable, and are precision engineered for purge-and-trap injection without cryofocusing. DB-MTBE columns operate effectively within temperature limits of 35 to 260/280 °C.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty