Would you like to log in again?
VariTide RPC Columns, Agilent Technologies
Supplier: Agilent Technologies
PL1512-5A05 PL1512-5A05
AGPL1512-5A05EA 882 USD InStock
AGPL1512-5A05 CAAGPL15125A05
VariTide RPC Columns, Agilent Technologies
Chromatography Columns
Agilent now offers VariTide RPC Columns. Please visit this VariTide RPC Columns page to learn more about VariTide RPC Columns.
  • A single column to cover the full range of synthetic peptides
  • Small particle size for maximum efficiency, even with 1 and 2 in prep columns
  • Bulk media to pack 1 and 2 in prep columns for the purification of mg to g quantities
The VariTide RPC columns and media are part of the VariPep Peptide Solution. This is the recommended option for cost-effective separation and purification of synthetic peptides using generic methods. The 200 Å pore size and 6 µm particle size provides a high efficiency purification solution for the full range of synthetic peptides.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty