Would you like to log in again?
J&W HP-PLOT Al₂O₃ M Columns, Agilent Technologies
Supplier: Agilent Technologies
19095P-M25PT 19095P-M23 19095P-M23 19091P-M15 19095P-M25PT 19091P-M15 19095P-M25 19095P-M25
AG19091P-M15EA 1459 USD InStock
CAAG19091P-M15 AG19091P-M15 AG19095P-M25 CAAG19095PM25P CAAG19095P-M23 AG19095P-M25PT AG19095P-M23 CAAG19095P-M25
J&W HP-PLOT Al₂O₃ M Columns, Agilent Technologies
Chromatography Columns
Agilent J&W HP-PLOT AL₂O₃ M is the most polar aluminum oxide phase deactivated with proprietary deactivation.
  • Each GC column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Similar phases are, AB-PLOT Al₂O₃ M, BGB-PLOT Al₂O₃ M and AT-Alumina
For details about the QC test conditions, including the test probe mixture and the resulting chromatogram, please refer to the Agilent J&W GC Column Performance Summary shipped with each GC column.
It is a good general use column for analysis of C1-C8 hydrocarbon isomers. It is excellent for resolving acetylene from butane and propylene from isobutene.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty