Would you like to log in again?
CF® Dye cAMP, Biotium
Supplier: Biotium
00036 00037
10018-140EA 139.91 USD InStock
10018-140 10018-142
CF® Dye cAMP, Biotium
Protein Linking and Labeling Systems
Fluorescently labeled cAMP analogs that can be used to probe cAMP receptors.
  • Superior CF® dyes are bright, photostable and water-soluble
  • Available with green or far-red labels
  • Soluble in H₂O, DMF, or DMSO
CF®488A-cAMP and CF®640R-cAMP are fluorescently labeled cAMP analogs that can be used to probe cAMP receptors.
Biotium’s next-generation CF® Dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
CF is a registered trademark of Biotium, Inc. Alexa Fluor, Texas Red, and DyLight are registered trademarks of Thermo Fisher Scientific.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty